Assay Details
Gene Symbol: LOC101102156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012486.3 | XP_004012535.2 | 5 - 6 | 545 | 66 | |
XM_027975244.1 | XP_027831045.1 | 5 - 6 | 545 | 66 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
