Assay Details
| Gene Symbol: PNLIPRP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012102992.3 | XP_011958382.1 | 2 - 3 | 333 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PNLIPRP3| Gene Symbol: PNLIPRP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012102992.3 | XP_011958382.1 | 2 - 3 | 333 | 92 | |
Set Membership: |
|