Assay Details
Gene Symbol: PNLIPRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012102992.3 | XP_011958382.1 | 7 - 8 | 941 | 121 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
