Assay Details
Gene Symbol: TNFSF13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973780.1 | XP_027829581.1 | 3 - 4 | 672 | 63 | |
XM_027973781.1 | XP_027829582.1 | 3 - 4 | 669 | 63 | ||
XM_027973782.1 | XP_027829583.1 | 3 - 4 | 672 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
