Assay Details
Gene Symbol: LOC114112797 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963906.1 | XP_027819707.1 | 1 | 154 | 104 |
Gene Symbol: LOC101113604 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973638.1 | XP_027829439.1 | 2 - 3 | 207 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
