Assay Details
| Gene Symbol: LOC101103111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960952.1 | XP_027816753.1 | 8 - 9 | 1094 | 95 | |
| Gene Symbol: LOC114110472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960951.1 | XP_027816752.1 | 5 - 6 | 554 | 95 | |
More Information
Set Membership: |
|

LOC101103111
Genomic Map