Assay Details
Gene Symbol: ALG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012166271.3 | XP_012021661.2 | 7 - 8 | 765 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
