Assay Details
| Gene Symbol: HAO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002387.4 | XP_004002436.2 | 3 - 4 | 584 | 76 | |
| XM_012179575.3 | XP_012034965.2 | 3 - 4 | 561 | 76 | ||
| XM_012179579.3 | XP_012034969.2 | 4 - 5 | 667 | 76 | ||
More Information
Set Membership: |
|

HAO2
Genomic Map