Assay Details
Gene Symbol: LOC101110257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976476.1 | XP_027832277.1 | 6 - 7 | 1112 | 65 |
Gene Symbol: LOC114117537 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976464.1 | XP_027832265.1 | 3 - 4 | 293 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
