Assay Details
Gene Symbol: CMTM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015047.4 | XP_004015096.1 | 2 - 3 | 423 | 68 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
