Assay Details
Gene Symbol: LSM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017216.4 | XP_004017265.1 | 2 - 3 | 222 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
