Assay Details
Gene Symbol: LOC101111335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004020949.4 | XP_004020998.2 | 2 - 3 | 294 | 92 | |
XM_027961899.1 | XP_027817700.1 | 2 - 3 | 294 | 92 | ||
XM_027961900.1 | XP_027817701.1 | 2 - 3 | 294 | 92 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
