Assay Details
| Gene Symbol: LOC101111335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020949.4 | XP_004020998.2 | 7 - 8 | 1149 | 73 | |
| XM_027961899.1 | XP_027817700.1 | 7 - 8 | 1149 | 73 | ||
| XM_027961900.1 | XP_027817701.1 | 7 - 8 | 1149 | 73 | ||
More Information
Set Membership: |
|

LOC101111335
Genomic Map