Assay Details
| Gene Symbol: LOC101112784 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020955.4 | XP_004021004.1 | 12 - 13 | 1128 | 112 | |
| XM_027962096.1 | XP_027817897.1 | 12 - 13 | 1316 | 112 | ||
More Information
Set Membership: |
|

LOC101112784
Genomic Map