Assay Details
Gene Symbol: LOC101112784 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004020955.4 | XP_004021004.1 | 6 - 7 | 754 | 96 | |
XM_027962096.1 | XP_027817897.1 | 6 - 7 | 966 | 96 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
