Assay Details
| Gene Symbol: FKBP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015099799.2 | XP_014955285.2 | 3 - 4 | 411 | 56 | |
| XM_027977025.1 | XP_027832826.1 | 3 - 4 | 411 | 56 | ||
More Information
Set Membership: |
|

FKBP1A
Genomic Map