Assay Details
Gene Symbol: CCDC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003518.4 | XP_004003567.2 | 7 - 8 | 1146 | 57 | |
XM_027968984.1 | XP_027824785.1 | 7 - 8 | 1144 | 57 | ||
XM_027968989.1 | XP_027824790.1 | 7 - 8 | 1133 | 57 | ||
XM_027968997.1 | XP_027824798.1 | 6 - 7 | 482 | 57 | ||
XM_027969003.1 | XP_027824804.1 | 5 - 6 | 605 | 57 | ||
XM_027969010.1 | XP_027824811.1 | 7 - 8 | 1194 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
