Assay Details
| Gene Symbol: LOC101113728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004431.2 | XP_004004480.1 | 2 - 3 | 305 | 65 | |
| XM_012128319.2 | XP_011983709.1 | 3 - 4 | 278 | 65 | ||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 LOC101113728
LOC101113728| Gene Symbol: LOC101113728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004431.2 | XP_004004480.1 | 2 - 3 | 305 | 65 | |
| XM_012128319.2 | XP_011983709.1 | 3 - 4 | 278 | 65 | ||
 Back To Top
 Back To Top