Assay Details
| Gene Symbol: EIF3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020865.4 | XP_004020914.1 | 11 - 12 | 1316 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EIF3C| Gene Symbol: EIF3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020865.4 | XP_004020914.1 | 11 - 12 | 1316 | 61 | |
Set Membership: |
|