Assay Details
| Gene Symbol: TMEM185B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012140208.3 | XP_011995598.2 | 1 | 541 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM185B| Gene Symbol: TMEM185B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012140208.3 | XP_011995598.2 | 1 | 541 | 101 | |
Set Membership: |
|