Assay Details
Gene Symbol: LOC101108654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004009524.3 | XP_004009573.1 | 1 - 2 | 109 | 100 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
