Assay Details
| Gene Symbol: APOC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015313.4 | XP_004015362.2 | 1 - 2 | 135 | 64 | |
| XM_027978463.1 | XP_027834264.1 | 1 - 2 | 135 | 64 | ||
| Gene Symbol: APOC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978462.1 | XP_027834263.1 | 1 - 2 | 108 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027978462.1 transcripts.
Set Membership: |
|

APOC4
Genomic Map