Assay Details
Gene Symbol: RPP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004023408.4 | XP_004023457.3 | 1 | 298 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
