Assay Details
| Gene Symbol: DNAJC19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012098384.3 | XP_011953774.2 | 2 - 3 | 184 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNAJC19| Gene Symbol: DNAJC19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012098384.3 | XP_011953774.2 | 2 - 3 | 184 | 76 | |
Set Membership: |
|