Assay Details
| Gene Symbol: BCAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978454.1 | XP_027834255.1 | 5 - 6 | 616 | 47 | |
| XM_027978455.1 | XP_027834256.1 | 5 - 6 | 706 | 47 | ||
More Information
Set Membership: |
|

BCAM
Genomic Map