Assay Details
Gene Symbol: LOC101119354 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002909.4 | XP_004002958.2 | 3 - 4 | 445 | 88 | |
XM_027979919.1 | XP_027835720.1 | 3 - 4 | 453 | 88 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
