Assay Details
Gene Symbol: MALL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006179.4 | XP_004006228.2 | 1 - 2 | 323 | 68 | |
XM_027967115.1 | XP_027822916.1 | 1 - 2 | 331 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
