Assay Details
Gene Symbol: KLHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012176.4 | XP_004012225.1 | 9 - 10 | 2022 | 68 | |
XM_027973689.1 | XP_027829490.1 | 8 - 9 | 1839 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
