Assay Details
Gene Symbol: LOC101114075 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004020871.4 | XP_004020920.1 | 6 - 7 | 1237 | 70 | |
XM_012104340.3 | XP_011959730.1 | 6 - 7 | 1273 | 70 | ||
XM_012104341.3 | XP_011959731.1 | 5 - 6 | 1026 | 70 | ||
XM_012104342.3 | XP_011959732.1 | 6 - 7 | 1233 | 70 | ||
XM_012104344.3 | XP_011959734.1 | 5 - 6 | 1062 | 70 | ||
XM_012104345.3 | XP_011959735.1 | 5 - 6 | 1022 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
