Assay Details
Gene Symbol: LOC101122140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012099260.2 | XP_011954650.1 | 3 - 4 | 1460 | 79 | |
XM_015102191.2 | XP_014957677.1 | 4 - 5 | 266 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
