Assay Details
Gene Symbol: ZFAND4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012105551.3 | XP_011960941.1 | 4 - 5 | 1209 | 60 | |
XM_012105552.2 | XP_011960942.1 | 4 - 5 | 532 | 60 | ||
XM_012105553.2 | XP_011960943.1 | 4 - 5 | 534 | 60 | ||
XM_012105556.3 | XP_011960946.1 | 4 - 5 | 1210 | 60 | ||
XM_012105562.2 | XP_011960952.1 | 5 - 6 | 503 | 60 | ||
XM_012105564.3 | XP_011960954.1 | 4 - 5 | 1223 | 60 | ||
XM_015104503.2 | XP_014959989.1 | 4 - 5 | 1218 | 60 | ||
XM_027962452.1 | XP_027818253.1 | 4 - 5 | 1271 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
