Assay Details
| Gene Symbol: TRIM50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961437.1 | XP_027817238.1 | 2 - 3 | 495 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRIM50| Gene Symbol: TRIM50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961437.1 | XP_027817238.1 | 2 - 3 | 495 | 65 | |
Set Membership: |
|