Assay Details
Gene Symbol: PTPN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976032.1 | XP_027831833.1 | 2 - 3 | 369 | 59 | |
XM_027976033.1 | XP_027831834.1 | 2 - 3 | 370 | 59 | ||
XM_027976034.1 | XP_027831835.1 | 2 - 3 | 369 | 59 | ||
XM_027976035.1 | XP_027831836.1 | 2 - 3 | 370 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
