Assay Details
Gene Symbol: RBM8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973971.1 | XP_027829772.1 | 5 - 6 | 1170 | 73 | |
XM_027973979.1 | XP_027829780.1 | 5 - 6 | 660 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
