Assay Details
Gene Symbol: POPDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011240.4 | XP_004011289.1 | 2 - 3 | 941 | 68 | |
XM_012182709.3 | XP_012038099.1 | 2 - 3 | 750 | 68 | ||
XM_012182710.3 | XP_012038100.1 | 2 - 3 | 788 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
