Assay Details
Gene Symbol: CRPPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027968498.1 | XP_027824299.1 | 1 - 2 | 271 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
