Assay Details
| Gene Symbol: LOC101103771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002522.3 | XP_004002571.2 | 2 - 3 | 260 | 83 | |
| XM_012181782.2 | XP_012037172.2 | 2 - 3 | 245 | 83 | ||
More Information
Set Membership: |
|

LOC101103771
Genomic Map