Assay Details
Gene Symbol: IL23R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003558.3 | XP_004003607.2 | 5 - 6 | 727 | 77 | |
XM_027964619.1 | XP_027820420.1 | 5 - 6 | 757 | 77 | ||
XM_027964628.1 | XP_027820429.1 | 5 - 6 | 727 | 77 | ||
GenBank mRNA | LN868336.1 | - | 5 - 6 | 728 | 77 | |
LN868337.1 | - | 5 - 6 | 758 | 77 | ||
LN868338.1 | - | 5 - 6 | 707 | 77 | ||
LN868339.1 | - | 5 - 6 | 728 | 77 | ||
LN868340.1 | - | 5 - 6 | 728 | 77 | ||
LN868341.1 | - | 5 - 6 | 728 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
