Assay Details
| Gene Symbol: ENDOG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005593.4 | XP_004005642.1 | 1 - 2 | 888 | 68 | |
| GenBank mRNA | AY221463.1 | - | 1 - 2 | 107 | 68 | |
More Information
Set Membership: |
|

ENDOG
Genomic Map