Assay Details
| Gene Symbol: ENDOG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005593.4 | XP_004005642.1 | 2 - 3 | 1000 | 91 | |
| GenBank mRNA | AY221463.1 | - | 2 - 3 | 219 | 91 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ENDOG| Gene Symbol: ENDOG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005593.4 | XP_004005642.1 | 2 - 3 | 1000 | 91 | |
| GenBank mRNA | AY221463.1 | - | 2 - 3 | 219 | 91 | |