Assay Details
| Gene Symbol: CYP24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976458.1 | XP_027832259.1 | 8 - 9 | 1154 | 93 | |
| XM_027976459.1 | XP_027832260.1 | 8 - 9 | 1154 | 93 | ||
More Information
Set Membership: |
|

CYP24
Genomic Map