Assay Details
| Gene Symbol: CYP24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976458.1 | XP_027832259.1 | 9 - 10 | 1234 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYP24| Gene Symbol: CYP24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976458.1 | XP_027832259.1 | 9 - 10 | 1234 | 112 | |
Set Membership: |
|