Assay Details
| Gene Symbol: MEAK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014942.4 | XP_004014991.2 | 2 - 3 | 260 | 81 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

MEAK7| Gene Symbol: MEAK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014942.4 | XP_004014991.2 | 2 - 3 | 260 | 81 | |