Assay Details
Gene Symbol: MEAK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014942.4 | XP_004014991.2 | 3 - 4 | 487 | 63 | |
XM_027977687.1 | XP_027833488.1 | 2 - 3 | 485 | 63 | ||
XM_027977688.1 | XP_027833489.1 | 2 - 3 | 444 | 63 |
