Assay Details
| Gene Symbol: MEAK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014942.4 | XP_004014991.2 | 7 - 8 | 1364 | 104 | |
| XM_027977687.1 | XP_027833488.1 | 6 - 7 | 1362 | 104 | ||
| XM_027977688.1 | XP_027833489.1 | 6 - 7 | 1321 | 104 | ||
More Information
Set Membership: |
|

MEAK7
Genomic Map