Assay Details
| Gene Symbol: LOC101103048 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004022703.4 | XP_004022752.3 | 1 | 1231 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101103048| Gene Symbol: LOC101103048 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004022703.4 | XP_004022752.3 | 1 | 1231 | 108 | |
Set Membership: |
|