Assay Details
| Gene Symbol: NDUFV2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015103655.2 | XP_014959141.2 | 4 - 5 | 389 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NDUFV2| Gene Symbol: NDUFV2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015103655.2 | XP_014959141.2 | 4 - 5 | 389 | 67 | |
Set Membership: |
|