Assay Details
| Gene Symbol: RWDD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972260.1 | XP_027828061.1 | 1 - 2 | 267 | 69 | |
| XM_027972270.1 | XP_027828071.1 | 1 - 2 | 282 | 69 | ||
More Information
Set Membership: |
|

RWDD3
Genomic Map