Assay Details
| Gene Symbol: C11H17orf100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975100.1 | XP_027830901.1 | 1 | 538 | 78 | |
| XM_027975101.1 | XP_027830902.1 | 1 | 538 | 78 | ||
More Information
Set Membership: |
|

C11H17orf100
Genomic Map