Assay Details
| Gene Symbol: C11H17orf100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975100.1 | XP_027830901.1 | 1 - 2 | 963 | 98 | |
| XM_027975101.1 | XP_027830902.1 | 1 - 2 | 963 | 98 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C11H17orf100| Gene Symbol: C11H17orf100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975100.1 | XP_027830901.1 | 1 - 2 | 963 | 98 | |
| XM_027975101.1 | XP_027830902.1 | 1 - 2 | 963 | 98 | ||